Learn More
Sino Biological Recombinant Human E-Cadherin Protein (hFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10204H02HB20
Encompass_Preferred
A DNA sequence encoding the human E-Cad (P12830-1)(Met1-Ile707) was expressed with the Fc region of human IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5432