Learn More
Sino Biological Recombinant Human Cadherin-11 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10065H08H1
Encompass_Preferred
A DNA sequence encoding the Human CDH11 (NP_001788) (Gly54-Thr617) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-5508