Learn More
Sino Biological Anti-Cadherin-16 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10915MM08100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human KSP-Cadherin / Cadherin-16 (rh KSP-Cadherin / Cadherin-16; Catalog#10915-H08H; NP_004053.1; Met1-Ala786). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1907