Learn More
Sino Biological Anti-Cadherin-16 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10915T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human KSP-Cadherin / Cadherin-16 / CDH16 (rh KSP-Cadherin / Cadherin-16 / CDH16; Catalog#10915-H08H; NP_004053.1; Met1-Ala786). KSP-Cadherin / Cadherin-16 / CDH16 specific IgG was purified by Human KSP-Cadherin / Cadherin-16 / CDH16 affinity chromatography.
Catalog No. 50-241-1731