Learn More
Sino Biological Human CDH17 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11360MM0220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CDH17 / Cadherin-17 (rh CDH17 / Cadherin-17; Catalog #11360-H08H; Met 1-Met 787; Q12864-1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3862