Learn More
Sino Biological Human CDH17 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11360MM0520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cadherin-17 / LI-cadherin / CDH17 (rh Cadherin-17 / LI-cadherin / CDH17; Catalog#11360-H08H; Q12864-1; Met1-Met787). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1148