Learn More
Sino Biological Recombinant Anti-N-Cadherin Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11039R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human N-Cadherin / CD325 / CDH2 (rh N-Cadherin / CD325 / CDH2; Catalog#11039-H08H; NP_001783.2; Met 1-Ala 724).
Catalog No. 50-240-7933