Learn More
Sino Biological Recombinant Anti-N-Cadherin Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 11039R001P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human N-Cadherin / CD325 / CDH2 (rh N-Cadherin / CD325 / CDH2; Catalog#11039-H08H; NP_001783.2; Met1-Ala724) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-2039