Learn More
Sino Biological Recombinant Human N-Cadherin Protein (His Tag)
Supplier:  Sino Biological 11039H08H21
Encompass_Preferred
A DNA sequence encoding the Human CDH2 (NP_001783.2) (Asp160-Ala724) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-4960