Learn More
Sino Biological Human CDH5 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11039R02020
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human VE-Cadherin / CD144 / CDH5 (CAA56306.1; Met1-Gln593).
Catalog No. 50-237-4418