Learn More
BIORTUS USA INC Human CDK3/CyclinE2 Protein, 100UG
Catalog No. NC2953289
Encompass_Preferred
Gene Name: CDK3/CCNE2. Uniprot: Q00526/O96020. Construct: GST-3C-CDK3(1-305end)/Flag-His-TEV-Cyclin E2 (S2-H404 end). Expression Host: Insect cell. Purity: 80%.
Catalog No. NC2953289 Supplier BIORTUS USA INC Supplier No. BP470E224A
Gene Name: CDK3/CCNE2. Uniprot: Q00526/O96020. Construct: GST-3C-CDK3(1-305end)/Flag-His-TEV-Cyclin E2 (S2-H404 end). Expression Host: Insect cell. Purity: 80%.