Learn More
Sino Biological Anti-CDK1/CDC2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 109678T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CDK1, and purified by antigen affinity chromatography.
Catalog No. 50-255-5017