Learn More
Sino Biological Anti-CDK2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10624RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CDK2 (rh CDK2; Catalog#10624-H08B; NP_001789.2; Met 1-Leu 298). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2204