Learn More
Sino Biological Human CDK4 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10732T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CDK4 (rh CDK4; Catalog#10732-H09B; NP_000066.1; Met1-Glu303). CDK4 specific IgG was purified by Human CDK4 affinity chromatography.
Catalog No. 50-237-3766