Learn More
Sino Biological Human CDK4 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 100873T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human CDK4, and purified by antigen affinity chromatography.
Catalog No. 50-237-1577