Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CDK4 Recombinant Protein

Catalog No. PIRP75530
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP75530 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP75530 Supplier Invitrogen™ Supplier No. RP75530
Add to Cart
Add to Cart

Recombinant Protein

This product contains purified recombinant human CDK4 protein with an N-terminal hexahistidine tag. This product may be used as a positive control in many immunological assays.

CDK4/cyclin D3 serine/threonine kinase is dimeric protein complex. CDK4 is a member of the cyclin-dependent protein kinase family and is involved in the control of cell proliferation during the G1 phase of cell cycle. CDK4 forms a complex with the D-type cyclins and is inhibited by p16 (cyclin-dependent kinase inhibitor-2). CDK4 can mediate phosphorylation of the C-terminal region of Rb protein leading to an active transcriptional repression of E2F complex. CDC37 and HSP90 can preferentially associate with the fraction of CDK4 not bound to D-type cyclins. SMAD3 is a major physiologic substrate of the G1 cyclin-dependent kinases CDK4 and CDK2. Although CDK6 and CDK4 can both phosphorylate multiple residues in the Rb protein, they do so with different residue selectivity in vitro; CDK6 phosphorylates Thr821 while CDK4 phosphorylates Thr826 on Rb protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11802
Concentration 0.081 mg/mL
For Use With (Application) Control, Western Blot
Formulation 0.01 M Bis-tris propane with 0.1 M NaCl, 50% glycerol and no preservative; pH 9.0
Gene ID (Entrez) 1019
Molecular Weight (g/mol) 38 kDa
Name Human CDK4
pH Range 9.0
Quantity 10 μg
Source E. coli
Immunogen N/A
Gene Alias Cdk4; Cell division protein kinase 4; CMM3; Crk3; cyclin dependent kinase 4; cyclin-dependent kinase 4; MGC14458; PSK-J3; serine/threonine kinase; Unknown (protein for MGC:133903)
Common Name CDK4
Gene Symbol CDK4
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Protein Tag His-tag
Sequence Amino acids Met1- Glu303 containing an N-terminal His-tag
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.