Learn More
Sino Biological Anti-CDKN2A/p16INK4a Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101169T3820
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CDKN2A, and purified by antigen affinity chromatography.
Catalog No. 50-237-0858