Learn More
BIOHIPPO Human CEA Microsample
SDP
Catalog No. 5000314777
Encompass
Human CEA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CEA in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CEA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CEA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CEA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14777 Supplier BIOHIPPO Supplier No. BHE15212671
Human CEA Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CEA in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 053 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CEA Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CEA Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CEA biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color