Learn More
Sino Biological Human CEACAM-1/CD66a Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000786638
Encompass_Preferred
The Human CEACAM1 ELISA Pair Set is for the quantitative determination of Human CEACAM1.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CEACAM1 coated on a 96-well plate. Standards and samples are added to the wells, and any Human CEACAM1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human CEACAM1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CEACAM1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86638 Supplier Sino Biological Supplier No. SEKA108225
The Human CEACAM1 ELISA Pair Set is for the quantitative determination of Human CEACAM1.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CEACAM1 coated on a 96-well plate. Standards and samples are added to the wells, and any Human CEACAM1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human CEACAM1 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CEACAM1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.