Learn More
R&D Systems™ Human CEACAM-3/CD66d (NP_001806) VersaClone cDNA
Catalog No. RDC0952
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
RDC0952 10 μg
1 options
This item is not returnable. View return policy
Catalog No. RDC0952 Supplier R&D Systems™ Supplier No. RDC0952
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Carcinoembryonic antigen-related cell adhesion molecule 3
The Human CEACAM-3/CD66d (NP_001806) VersaClone cDNA clone contains the complete ORF for CEACAM3/CD66d, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CEACAM3/CD66d insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
Specifications
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Quantity 10 μg
For Reasearch Use Only; Not for Use in Humans