Learn More
ACROBiosystems Human CEACAM-5 100ug
Supplier:  ACROBiosystems CE5H52H8100UG
Encompass_Preferred
Human CEACAM-5 (499-685) Protein His Tag (CE5-H52H8) is expressed from human 293 cells (HEK293). It contains AA Glu 499 - Ala 685 (Accession # P06731-1).
Catalog No. 50-253-5305