Learn More
Sino Biological Human CEACAM-5/CD66e Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785936
Encompass_Preferred
The Human CEACAM5 / CD66e ELISA Pair Set is for the quantitative determination of Human CEACAM5 / CD66e.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CEACAM5 / CD66e coated on a 96-well plate. Standards and samples are added to the wells, and any Human CEACAM5 / CD66e present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human CEACAM5 / CD66e monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CEACAM5 / CD66e present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85936 Supplier Sino Biological Supplier No. SEK110775
The Human CEACAM5 / CD66e ELISA Pair Set is for the quantitative determination of Human CEACAM5 / CD66e.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CEACAM5 / CD66e coated on a 96-well plate. Standards and samples are added to the wells, and any Human CEACAM5 / CD66e present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human CEACAM5 / CD66e monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CEACAM5 / CD66e present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.