Learn More
Sino Biological Human CEACAM-6/CD66c Matched ELISA Antibody Pair Set 5 Plates
Supplier:  Sino Biological SEKA108235
Encompass_Preferred
The Human CEACAM6 / CD66c ELISA Pair Set is for the quantitative determination of Human CEACAM6 / CD66c.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human CEACAM6 / CD66c coated on a 96-well plate. Standards and samples are added to the wells, and any Human CEACAM6 / CD66c present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human CEACAM6 / CD66c monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human CEACAM6 / CD66c present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86250