Learn More
Bon Opus Biosciences Recombinant Human CEACAM1 (C-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP107150UG
Encompass_Preferred
Recombinant Human CEACAM1 (C-His) is produced by Human Cells. The target gene encoding Gln35-Gly428 is expressed with a C-His tag. Accession# P13688. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1071
Catalog No. 50-217-2669