Learn More
Sino Biological Recombinant Anti-CEACAM-1/CD66a Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10822R006100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CEACAM1 / CD66a (rh CEACAM1 / CD66a; Catalog#10822-H08H; NP_001020083.1; Met 1-Gly 428).
Catalog No. 50-240-9238