Learn More
Sino Biological Anti-CEACAM-1/CD66a Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10822MM02P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CEACAM1 / CD66a (rh CEACAM1 / CD66a; Catalog#10822-H08H; NP_001020083.1; Met1-Gly428) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-8214