Learn More
Sino Biological Human CEACAM1 MAb 25TS (Sino Biological)
Supplier:  Sino Biological 10822R103P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CEACAM1 / CD66a (rh CEACAM1 / CD66a; Catalog#10822-H08H; NP_001020083.1; Met1-Gly428) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-1044