Learn More
Sino Biological Human CEACAM1 MAb 100TS (Sino Biological)
Supplier:  Sino Biological 10822MM02F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CEACAM1 / CD66a (rh CEACAM1 / CD66a; Catalog#10822-H08H; NP_001020083.1; Met1-Gly428) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-237-2445