Learn More
Sino Biological Human CEACAM5 MAb 100ug (Sino Biological)
Supplier:  Sino Biological 11077MM04H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CEACAM5 / CEA / CD66e (rh CEACAM5 / CEA / CD66e; Catalog#11077-H08H; NP_004354.2; Met1-Ala685). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-237-2632