Learn More
Sino Biological Anti-CEACAM-5/CD66e Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11077RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CEACAM5 (rh CEACAM5; Catalog#11077-H08H; NP_004354.2; Met 1-Ala 685). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4874