Learn More
Sino Biological Recombinant Human CEACAM-1/CD66a Protein (ECD, His Tag), HPLC-verified 1 mg
Supplier:  Sino Biological 10822H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CEACAM1 (NP_001020083.1) (Met 1-Gly 428) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-222-3568