Learn More
Sino Biological Anti-CEACAM-6/CD66c Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10823MM12A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CEACAM6 / CD66c (rh CEACAM6 / CD66c; Catalog#10823-H08H; NP_002474.4; Met1-Gly320) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-240-9781