Learn More
Sino Biological Recombinant Anti-CEACAM-6/CD66c Antibody (PerCP), Rabbit Monoclonal
Supplier:  Sino Biological 10823R408C25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CEACAM6 / CD66c (rh CEACAM6 / CD66c; 10823-H08H; Met 1-Gly 320; NP_002474.4) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-241-1400