Learn More
Sino Biological Human CEACAM6 MAb 100TS (Sino Biological)
Supplier:  Sino Biological 10823R408F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CEACAM6 / CD66c (rh CEACAM6 / CD66c; 10823-H08H; Met 1-Gly 320; NP_002474.4) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-237-0755