Learn More
Sino Biological Recombinant Human CEACAM-6/CD66c Protein (His Tag), HPLC-verified, Amine-Labeled Biotinylated
Supplier:  Sino Biological 10823H08HB100
Encompass_Preferred
A DNA sequence encoding the human CEACAM6 (NP_002474.3) (Met1-Gly320) with a C-terminal polyhistidine tag was expressed. The purified protein was biotinylated in vitro.
Catalog No. 50-255-5150