Learn More
Sino Biological Human CEACAM6 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 10823H49HB100
Encompass_Preferred
A DNA sequence encoding the Human CEACAM6 (NP_002474.3) (Met1-Gly320) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-2454