Learn More
Sino Biological Human CEACAM6 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 10823H02H1
Encompass_Preferred
A DNA sequence encoding the Human CEACAM6 (NP_002474.3) (Met1-Gly320) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2449