Learn More
Sino Biological Recombinant Human CEACAM-8/CD66b Protein (His & AVI Tag), Biotinylated
Supplier:  Sino Biological 11729H27HB100
Encompass_Preferred
A DNA sequence encoding the human CEACAM8 (NP_001807.2) (Met1-Ser319) was expressed with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4597