Learn More
Biomatik Corporation Human Cellular Repressor Of E1A Stimulated Genes 1 (CREG1) ELISA Kit
Supplier:  Biomatik Corporation EKU03067
Encompass_Preferred
Human Cellular Repressor Of E1A Stimulated Genes 1 (CREG1) ELISA Kit for detecting CREG1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.123ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75629. Gene ID: 8804. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5052