Learn More
BIOHIPPO Human CENPF Microsample
SDP
Catalog No. 5000315509
Encompass
Human CENPF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CENPF in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CENPF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CENPF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CENPF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15509 Supplier BIOHIPPO Supplier No. BHE15211806
Human CENPF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CENPF in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 054 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CENPF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CENPF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CENPF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color