Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CENTB1 (aa 205-274) Control Fragment Recombinant Protein

Catalog No. RP107134
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP107134 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP107134 Supplier Invitrogen™ Supplier No. RP107134
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (90%), Rat (90%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84987. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

ACAP1, also known as Centaurin-beta-1 (CENTB1), is a 740 amino acid containing transport carrier belonging to the Centaurin family with three ANK repeats, an Arf-GAP domain, a BAR domain and a PH domain that binds to phospholipids. ACAP1, a GTPase-activating protein (GAP) for ADP ribosylation factor 6 (ARF6), is required for clathrin-dependent export of proteins from recycling endosomes to trans-Golgi network and cell surface. It mediates CENTB1-binding to phosphatidylinositol 3,5-bisphosphate (PIP2) or phosphatidylinositol 3,4,5-trisphosphate (PIP3) containing membranes. Along with ACAP2, ACAP1 is recruited to platelet-derived growth factor (PDGF) induced dorsal membrane ruffles in NIH 3T3 mouse fibroblasts, while over expression inhibited ruffle formation. It is widely expressed in most tissues with high expression in lung and spleen and low expression in heart, kidney, liver and pancreas.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q15027
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 9744
Name Human CENTB1 (aa 205-274) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Acap1; Arf GAP with coiled coil, ANK repeat and PH domains 1; arf-GAP with coiled-coil, ANK repeat and PH domain-containing protein 1; ArfGAP with coiled-coil, ankyrin repeat and PH domains 1; centaurin beta 1; centaurin, beta 1; centaurin-beta-1; Centb1; cnt-b1; KIAA0050
Common Name CENTB1
Gene Symbol ACAP1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence QGHEELSRLSQYRKELGAQLHQLVLNSAREKRDMEQRHVLLKQKELGGEEPEPSLREGPGGLVMEGHLFK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.