Learn More
Biomatik Corporation Human Centromere Protein F (CENPF) ELISA Kit
Supplier:  Biomatik Corporation EKU03072
Encompass_Preferred
Human Centromere Protein F (CENPF) ELISA Kit for detecting CENPF in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49454. Gene ID: 1063. Target Synonyms: CENP-F; CENF; Hcp-1; Centromere protein F (350/400kD,Mitosin) ; AH antigen; Kinetochore protein CENPF; Mitosin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5943