Learn More
Sino Biological Anti-CER1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10820T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CER1 / Cerberus 1 (rh CER1 / Cerberus 1; Catalog#10820-H08B; AAH69371.1; Met1-Ala267). CER1 / Cerberus 1 specific IgG was purified by Human CER1 / Cerberus 1 affinity chromatography.
Catalog No. 50-241-2021