Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Cerberus 1 ELISA Kit

Catalog No. EH102RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH102RBX5 5 x 96 Tests
EH102RB 96 Tests
EH102RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH102RBX5 Supplier Invitrogen™ Supplier No. EH102RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

ELISA

Human Cerberus 1 quantitates human Cerberus 1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cerberus 1.

This gene encodes a cytokine member of the cysteine knot superfamily, characterized by nine conserved cysteines and a cysteine knot region. The cerberus-related cytokines, together with Dan and DRM/Gremlin, represent a group of bone morphogenetic protein antagonists that can bind directly to BMPs and inhibit their activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O95813
Assay Range 0.82-200 ng/mL
Assay Sensitivity 0.82 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias Cer1; cer-1; Cerberus; cerberus 1 homolog (Xenopus laevis); cerberus 1, cysteine knot superfamily, homolog; cerberus 1, DAN family BMP antagonist; cerberus homolog; cerberus-like protein; cerberus-related 1; cerberus-related protein; Cerl; Cer-l; Cerl1; Cerr1; DAN domain family member 4; DAND4; MGC119894; MGC119895; MGC96951; RGD1563046Cer1; cer-1; Cerberus; cerberus 1 homolog (Xenopus laevis); cerberus 1, cysteine knot superfamily, homolog; cerberus 1, DAN family BMP antagonist; cerberus homolog; cerberus-like protein; cerberus-related 1; cerberus-related protein; Cerl; Cer-l; Cerl1; Cerr1; DAN domain family member 4; DAND4; MGC119894; MGC119895; MGC96951; RGD1563046;
Gene ID (Entrez) 9350
Gene Symbol CER1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target CER1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.