Learn More
Biomatik Corporation Human Cerberus (CER) CLIA Kit
Supplier:  Biomatik Corporation EKU09831
Encompass_Preferred
Human Cerberus (CER) CLIA Kit for detecting CER in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.21pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O95813. Gene ID: 9350. Target Synonyms: CER1; DAND4; Cerberus 1; DAN domain family member 4; Cysteine Knot Superfamily,Homolog (Xenopus Laevis) ; Cerberus-related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8423