Learn More
Biomatik Corporation Human Cerberus (CER) ELISA Kit
Supplier:  Biomatik Corporation EKU03074
Encompass_Preferred
Human Cerberus (CER) ELISA Kit for detecting CER in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 13.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O95813. Gene ID: 9350. Target Synonyms: CER1; DAND4; Cerberus 1; DAN domain family member 4; Cysteine Knot Superfamily,Homolog (Xenopus Laevis) ; Cerberus-related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6687