Learn More
Biomatik Corporation Human Cereblon (CRBN) ELISA Kit
Supplier:  Biomatik Corporation EKU03077
Encompass_Preferred
Human Cereblon (CRBN) ELISA Kit for detecting CRBN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96SW2. Gene ID: 51185. Target Synonyms: MRT2A; Mental Retardation,Non-Syndromic,Autosomal Recessive 2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9065