Learn More
Biomatik Corporation Human Ceruloplasmin (CP) CLIA Kit
Supplier:  Biomatik Corporation EKU09475
Encompass_Preferred
Human Ceruloplasmin (CP) CLIA Kit for detecting CP in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.58pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P00450. Gene ID: 1356. Target Synonyms: CP2; CER; Caeruloplasmin; Ferroxidase; Oxygen Oxidoreductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8512