Learn More
Sino Biological Recombinant Human carboxylesterase 2/CES2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10380H08H50
Encompass_Preferred
A DNA sequence encoding the human CES2 isoform 1 (O00748-1) (Met 1-Leu 559) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-224-7160