Learn More
Eagle Biosciences Inc Human cFGF-21 ELISA Assay Kit
SDP
Supplier:  Eagle Biosciences Inc CTF31K01
Encompass
This Eagle Biosciences Human C-terminal FGF-21 ELISA Assay Kit is a sandwich ELISA (enzyme-linked immunosorbent assay) and intended for the quantitative determination of human C-terminal FGF-21 level in EDTA-plasma or serum. It measures both the intact FGF-21 and the C-terminal FGF-21 fragments that must not be C-terminally truncated while the N-terminal end of the FGF-21 may be truncated. The test is useful in clinical study related to diabetes and obesity, and is for research use only, not for use in diagnostic procedures.
Catalog No. 50-187-8338